missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (Chloromethyl)phosphonate 98.0+%, TCI America™

Catalog No. d45881g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45881G 1 g
D45885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45881G Supplier TCI America Supplier No. D45881G
Only null left

Chemical Identifiers

CAS 3167-63-3
Molecular Formula C5H12ClO3P
Molecular Weight (g/mol) 186.57
MDL Number MFCD00010189
InChI Key MZBIWKMCTWJLPT-UHFFFAOYSA-N
Synonym diethyl chloromethyl phosphonate, diethyl chloromethylphosphonate, diethyl chloromethanephosphonate, phosphonic acid, chloromethyl-, diethyl ester, chloromethylphosphonic acid diethyl ester, phosphonic acid, p-chloromethyl-, diethyl ester, chloromethyl phosphonic acid diethyl ester, 1-chloromethyl ethoxy phosphoryl oxyethane, acmc-1cq3s
PubChem CID 76633
IUPAC Name diethyl (chloromethyl)phosphonate
SMILES CCOP(=O)(CCl)OCC

Specifications

Color Yellow
Boiling Point 110°C
Quantity 1 g
Formula Weight 186.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (Chloromethyl)phosphonate