missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Carbonate 98.0+%, TCI America™

Catalog No. p-7124191
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
C004125G 25 g
C0041500G 500 g
2 options

Catalog No. C004125G

Supplier: TCI America C004125G

Only null left

Chemical Identifiers

CAS 105-58-8
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00009107
InChI Key OIFBSDVPJOWBCH-UHFFFAOYSA-N
Synonym ethyl carbonate, carbonic acid, diethyl ester, carbonic acid diethyl ester, eufin, diatol, diaethylcarbonat, carbonic ether, ethoxyformic anhydride, diethylkarbonat, ethyl carbonate eto 2co
PubChem CID 7766
IUPAC Name diethyl carbonate
SMILES CCOC(=O)OCC

Specifications

Color Colorless
Boiling Point 126°C
Quantity 25 g
UN Number 2366
Formula Weight 118.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Carbonate
EINECSNumber (2)-1699
RTECSNumber FF9800000
TSCA Yes