missing translation for 'onlineSavingsMsg'
Learn More

Diethyl carbonate, 99%

Catalog No. AC114225000
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114225000 500 mL Glass bottle
AC114221000 100 mL Glass bottle
AC114220010 1 L Glass bottle
AC114220025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC114225000 Supplier Thermo Scientific Chemicals Supplier No. 114225000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 105-58-8 | C5H10O3 | 118.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Alkylation reagent

Chemical Identifiers

CAS 105-58-8
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00009107
InChI Key OIFBSDVPJOWBCH-UHFFFAOYSA-N
Synonym ethyl carbonate, carbonic acid, diethyl ester, carbonic acid diethyl ester, eufin, diatol, diaethylcarbonat, carbonic ether, ethoxyformic anhydride, diethylkarbonat, ethyl carbonate eto 2co
PubChem CID 7766
IUPAC Name diethyl carbonate
SMILES CCOC(=O)OCC

Specifications

Melting Point -43.0°C
Density 0.9750g/mL
Boiling Point 126.0°C to 128.0°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3830 to 1.3850
Linear Formula (C2H5O)2CO
Quantity 500 mL
Beilstein 03,5
Fieser 01,247; 02,129; 03,95; 05,213
Merck Index 15,3835
Specific Gravity 0.975
Solubility Information Solubility in water: 18.8g/L
Viscosity 0.827 mPa.s (20°C)
Formula Weight 118.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethyl carbonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-311-1

RUO – Research Use Only