missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Bromomalonate 85.0+%, TCI America™

Catalog No. p-7118920
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
B120525G 25 g
B1205500G 500 g
2 options

Catalog No. B120525G

Supplier: TCI America B120525G

Only null left

Chemical Identifiers

CAS 685-87-0
Molecular Formula C7H11BrO4
Molecular Weight (g/mol) 239.065
MDL Number MFCD00009138
InChI Key FNJVDWXUKLTFFL-UHFFFAOYSA-N
Synonym diethyl bromomalonate, diethyl 2-bromomalonate, bromomalonic acid diethyl ester, ethyl bromomalonate, diethylbromomalonate, propanedioic acid, bromo-, diethyl ester, 1,3-diethyl 2-bromopropanedioate, diethyl bromopropanedioate, malonic acid, bromo-, diethyl ester, propanedioic acid, 2-bromo-, 1,3-diethyl ester
PubChem CID 69637
IUPAC Name diethyl 2-bromopropanedioate
SMILES CCOC(=O)C(C(=O)OCC)Br

Specifications

Color Yellow
Boiling Point 235°C
Quantity 25 g
UN Number 3265
Formula Weight 239.07
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Bromomalonate