Learn More
Diethyl bromomalonate, 90+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 114191000
| Quantity | 100mL |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C7H11BrO4 | |
| MFCD00009138 | |
| diethyl bromomalonate, diethyl 2-bromomalonate, bromomalonic acid diethyl ester, ethyl bromomalonate, diethylbromomalonate, propanedioic acid, bromo-, diethyl ester, 1,3-diethyl 2-bromopropanedioate, diethyl bromopropanedioate, malonic acid, bromo-, diethyl ester, propanedioic acid, 2-bromo-, 1,3-diethyl ester | |
| diethyl 2-bromopropanedioate |
Specifications
| Diethyl bromomalonate | |
| 1.4000g/mL | |
| >110°C | |
| 90% min. (GC) | |
| C7H11BrO4 | |
| C2H5O2CCH(Br)CO2C2H5 | |
| MFCD00009138 | |
| 1.4 | |
| Solubility in water: immiscible | |
| CCOC(=O)C(C(=O)OCC)Br | |
| 239.07 | |
| 239.07 |
| 685-87-0 | |
| 233°C to 235°C | |
| Authentic | |
| Glass bottle | |
| 1.4520 to 1.456 | |
| 100mL | |
| 02, 594 | |
| diethyl bromomalonate, diethyl 2-bromomalonate, bromomalonic acid diethyl ester, ethyl bromomalonate, diethylbromomalonate, propanedioic acid, bromo-, diethyl ester, 1,3-diethyl 2-bromopropanedioate, diethyl bromopropanedioate, malonic acid, bromo-, diethyl ester, propanedioic acid, 2-bromo-, 1,3-diethyl ester | |
| FNJVDWXUKLTFFL-UHFFFAOYSA-N | |
| diethyl 2-bromopropanedioate | |
| 69637 | |
| 90+% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 211-683-1
Promotions