missing translation for 'onlineSavingsMsg'
Learn More

Diethyl bromomalonate, 90+%

Catalog No. p-7081133
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
Catalog No. Quantity
AAA1096514 25 g
AAA1096518 50 g
AAA1096530 250 g
3 options

Catalog No. AAA1096514

Supplier: Thermo Scientific Chemicals A1096514

Only null left

CAS: 685-87-0 | C7H11BrO4 | 239.065 g/mol

Diethyl bromomalonate is used in perfumes. It is also used to synthesize other compounds such as barbiturates, artificial flavorings, vitamin B1, and vitamin B6.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl bromomalonate is used in perfumes. It is also used to synthesize other compounds such as barbiturates, artificial flavorings, vitamin B1, and vitamin B6.

Solubility
Does not mix well with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store at room temperature. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 685-87-0
Molecular Formula C7H11BrO4
Molecular Weight (g/mol) 239.065
MDL Number MFCD00009138
InChI Key FNJVDWXUKLTFFL-UHFFFAOYSA-N
Synonym diethyl bromomalonate, diethyl 2-bromomalonate, bromomalonic acid diethyl ester, ethyl bromomalonate, diethylbromomalonate, propanedioic acid, bromo-, diethyl ester, 1,3-diethyl 2-bromopropanedioate, diethyl bromopropanedioate, malonic acid, bromo-, diethyl ester, propanedioic acid, 2-bromo-, 1,3-diethyl ester
PubChem CID 69637
IUPAC Name diethyl 2-bromopropanedioate
SMILES CCOC(=O)C(C(=O)OCC)Br

Specifications

Density 1.402
Boiling Point 233°C to 235°C
Flash Point 111°C (231°F)
Refractive Index 1.455
Quantity 25 g
UN Number UN3265
Beilstein 509768
Solubility Information Does not mix well with water.
Formula Weight 239.07
Percent Purity ≥90%
Chemical Name or Material Diethyl bromomalonate
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 211-683-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only