Learn More
Diethyl bis(hydroxymethyl)malonate, 95%
CAS: 20605-01-0 | C9H16O6 | 220.221 g/mol
Supplier: Thermo Scientific Chemicals A1367736
Description
Diethyl bis(hydroxymethyl)malonate is used as a reagent for preparing 1,3-dioxanes from acetals, aldehydes and ketones. It is also used as an intermediate for the preparation of substituted malonic, acrylic and isobutyric esters, for conversion to the useful dienophiles, ?-bromomethylacrylic acid and its ethyl ester and to bis(bromomethyl)acetic acid and its methyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl bis(hydroxymethyl)malonate is used as a reagent for preparing 1,3-dioxanes from acetals, aldehydes and ketones. It is also used as an intermediate for the preparation of substituted malonic, acrylic and isobutyric esters, for conversion to the useful dienophiles, É-bromomethylacrylic acid and its ethyl ester and to bis(bromomethyl)acetic acid and its methyl ester.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
| Diethyl bis(hydroxymethyl)malonate | |
| 47°C to 52°C | |
| C9H16O6 | |
| 500 g | |
| diethyl bis hydroxymethyl malonate, diethyl 2,2-bis hydroxymethyl malonate, propanedioic acid, bis hydroxymethyl-, diethyl ester, 1,3-diethyl 2,2-bis hydroxymethyl propanedioate, diethyl 2,2-bis hydroxymethyl propanedioate, 2,2-bis hydroxymethyl malonic acid diethyl ester, bis hydroxymethyl malonic acid diethyl ester, diethyl 2,2-bis hydroxymethyl propane-1,3-dioate, acmc-20al8x, ksc490a8h | |
| WIOHBOKEUIHYIC-UHFFFAOYSA-N | |
| diethyl 2,2-bis(hydroxymethyl)propanedioate | |
| 311844 | |
| 97% |
| 20605-01-0 | |
| >110°C (230°F) | |
| MFCD00009130 | |
| 1788515 | |
| Slightly soluble in water. | |
| CCOC(=O)C(CO)(CO)C(=O)OCC | |
| 220.221 | |
| 220.22 |
Chemical Identifiers
| 20605-01-0 | |
| 220.221 | |
| WIOHBOKEUIHYIC-UHFFFAOYSA-N | |
| 311844 | |
| CCOC(=O)C(CO)(CO)C(=O)OCC |
| C9H16O6 | |
| MFCD00009130 | |
| diethyl bis hydroxymethyl malonate, diethyl 2,2-bis hydroxymethyl malonate, propanedioic acid, bis hydroxymethyl-, diethyl ester, 1,3-diethyl 2,2-bis hydroxymethyl propanedioate, diethyl 2,2-bis hydroxymethyl propanedioate, 2,2-bis hydroxymethyl malonic acid diethyl ester, bis hydroxymethyl malonic acid diethyl ester, diethyl 2,2-bis hydroxymethyl propane-1,3-dioate, acmc-20al8x, ksc490a8h | |
| diethyl 2,2-bis(hydroxymethyl)propanedioate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 243-916-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only