missing translation for 'onlineSavingsMsg'
Learn More

Diethyl benzylphosphonate, 99%

Catalog No. AAA1064518
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1064518 50 g
AAA1064530 250 g
AAA106450B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1064518 Supplier Thermo Scientific Chemicals Supplier No. A1064518
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1080-32-6 | C11H17O3P | 228.228 g/mol

Diethyl benzylphosphonate is used as a reactant for synthesis of 3,5-dihydroxy-4-isopropylstilbene for treatment of skin disorders and natural cytotoxic marine products of polyketide origin via intramolecular Diels-Alder reactions. It is also used as reactant for cyclization of aryl ethers, amines and amides & for investigating the effects of functional groups on the performance of clue organic LEDs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl benzylphosphonate is used as a reactant for synthesis of 3,5-dihydroxy-4-isopropylstilbene for treatment of skin disorders and natural cytotoxic marine products of polyketide origin via intramolecular Diels-Alder reactions. It is also used as reactant for cyclization of aryl ethers, amines and amides & for investigating the effects of functional groups on the performance of clue organic LEDs.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1080-32-6
Molecular Formula C11H17O3P
Molecular Weight (g/mol) 228.228
MDL Number MFCD00009078
InChI Key AIPRAPZUGUTQKX-UHFFFAOYSA-N
Synonym diethyl benzylphosphonate, benzylphosphonic acid diethyl ester, diethoxyphosphonomethylbenzene, diethyl phosphonate, benzyl, phosphonic acid, phenylmethyl-, diethyl ester, unii-t6d4qwt656, diethylbenzylphosphonate, phosphonic acid, benzyl-, diethyl ester, diethyl phenylmethyl phosphonate, phosphonic acid, p-phenylmethyl-, diethyl ester
PubChem CID 14122
IUPAC Name diethoxyphosphorylmethylbenzene
SMILES CCOP(=O)(CC1=CC=CC=C1)OCC

Specifications

Density 1.094
Boiling Point 106°C to 108°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4975
Quantity 50 g
Beilstein 2580931
Solubility Information Insoluble in water.
Formula Weight 228.23
Percent Purity 99%
Chemical Name or Material Diethyl benzylphosphonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-097-4
RTECSNumber SZ6600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only