missing translation for 'onlineSavingsMsg'
Learn More

Diethyl benzylmalonate, 97%

Catalog No. p-7026720
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2289014 25 g
AAB2289022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2289014 Supplier Thermo Scientific Chemicals Supplier No. B2289014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 607-81-8 | C14H18O4 | 250.294 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 607-81-8
Molecular Formula C14H18O4
Molecular Weight (g/mol) 250.294
MDL Number MFCD00009166
InChI Key ICZLTZWATFXDLP-UHFFFAOYSA-N
Synonym diethyl benzylmalonate, diethyl 2-benzylmalonate, benzylmalonic acid diethyl ester, propanedioic acid, phenylmethyl-, diethyl ester, unii-40y0x0xn6p, malonic acid, benzyl-, diethyl ester, malonic acid, 2-benzyl-, diethyl ester, diethyl benzyl malonate, 1,3-diethyl 2-benzylpropanedioate, diethyl benzylpropanedioate
PubChem CID 69090
IUPAC Name diethyl 2-benzylpropanedioate
SMILES CCOC(=O)C(CC1=CC=CC=C1)C(=O)OCC

Specifications

Density 1.069
Boiling Point 162°C to 163°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.486
Quantity 25 g
Beilstein 615793
Formula Weight 250.29
Percent Purity 97%
Chemical Name or Material Diethyl benzylmalonate
EINECSNumber 210-142-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only