Learn More
Diethyl aminomalonate hydrochloride, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 13433-00-6 |
|---|---|
| Molecular Formula | C7H13NO4·ClH |
| Molecular Weight (g/mol) | 211.64 |
| MDL Number | MFCD00012510 |
| InChI Key | GLFVNTDRBTZJIY-UHFFFAOYSA-N |
| Synonym | diethyl aminomalonate hydrochloride, diethyl 2-aminomalonate hydrochloride, aminomalonic acid diethyl ester hydrochloride, 1,3-diethyl 2-aminopropanedioate hydrochloride, propanedioic acid, amino-, diethyl ester, hydrochloride, diethyl 2-aminopropanedioate hydrochloride, diethyl 2-aminomalonate hcl, diethyl aminomalonate hcl, diethyl 2-aminopropane-1,3-dioate, chloride, pubchem23977 |
| PubChem CID | 3084143 |
| IUPAC Name | diethyl 2-aminopropanedioate;hydrochloride |
| SMILES | CCOC(=O)C(C(=O)OCC)N.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC114040250
|
Thermo Scientific Chemicals
114040250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC114041000
|
Thermo Scientific Chemicals
114041000 |
100g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 13433-00-6 | |
| 211.64 | |
| GLFVNTDRBTZJIY-UHFFFAOYSA-N | |
| 3084143 | |
| CCOC(=O)C(C(=O)OCC)N.Cl |
| C7H13NO4·ClH | |
| MFCD00012510 | |
| diethyl aminomalonate hydrochloride, diethyl 2-aminomalonate hydrochloride, aminomalonic acid diethyl ester hydrochloride, 1,3-diethyl 2-aminopropanedioate hydrochloride, propanedioic acid, amino-, diethyl ester, hydrochloride, diethyl 2-aminopropanedioate hydrochloride, diethyl 2-aminomalonate hcl, diethyl aminomalonate hcl, diethyl 2-aminopropane-1,3-dioate, chloride, pubchem23977 | |
| diethyl 2-aminopropanedioate;hydrochloride |
Specifications
| 13433-00-6 | |
| 97.5% min. (Argentometry) | |
| C7H13NO4·ClH | |
| MFCD00012510 | |
| 04, 470 | |
| diethyl aminomalonate hydrochloride, diethyl 2-aminomalonate hydrochloride, aminomalonic acid diethyl ester hydrochloride, 1,3-diethyl 2-aminopropanedioate hydrochloride, propanedioic acid, amino-, diethyl ester, hydrochloride, diethyl 2-aminopropanedioate hydrochloride, diethyl 2-aminomalonate hcl, diethyl aminomalonate hcl, diethyl 2-aminopropane-1,3-dioate, chloride, pubchem23977 | |
| GLFVNTDRBTZJIY-UHFFFAOYSA-N | |
| diethyl 2-aminopropanedioate;hydrochloride | |
| 3084143 | |
| 98% |
| Authentic | |
| Glass bottle | |
| C2H5O2CCH(NH2)CO2C2H5·HCl | |
| 25g | |
| 15,132 | |
| Solubility in water: soluble | |
| CCOC(=O)C(C(=O)OCC)N.Cl | |
| 211.64 | |
| 211.64 | |
| Diethyl aminomalonate hydrochloride |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 236-556-8
RUO â Research Use Only