missing translation for 'onlineSavingsMsg'
Learn More

Diethyl aminomalonate hydrochloride, 98%

Catalog No. AAA1368130
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1368130 250 g
AAA1368109 10 g
AAA1368118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1368130 Supplier Thermo Scientific Chemicals Supplier No. A1368130
Only null left

CAS: 13433-00-6 | C7H14ClNO4 | 211.642 g/mol

Diethyl aminomalonate hydrochloride is used as a pharmaceutical intermediate. Diethyl aminomalonate with formaldehyde generates an azomethine ylide which can undergo cycloadditions with 1,2-dipolarophiles to form pyrrolidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl aminomalonate hydrochloride is used as a pharmaceutical intermediate. Diethyl aminomalonate with formaldehyde generates an azomethine ylide which can undergo cycloadditions with 1,2-dipolarophiles to form pyrrolidines.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is hygroscopic in nature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 13433-00-6
Molecular Formula C7H14ClNO4
Molecular Weight (g/mol) 211.642
MDL Number MFCD00012510
InChI Key GLFVNTDRBTZJIY-UHFFFAOYSA-N
Synonym diethyl aminomalonate hydrochloride, diethyl 2-aminomalonate hydrochloride, aminomalonic acid diethyl ester hydrochloride, 1,3-diethyl 2-aminopropanedioate hydrochloride, propanedioic acid, amino-, diethyl ester, hydrochloride, diethyl 2-aminopropanedioate hydrochloride, diethyl 2-aminomalonate hcl, diethyl aminomalonate hcl, diethyl 2-aminopropane-1,3-dioate, chloride, pubchem23977
PubChem CID 3084143
IUPAC Name diethyl 2-aminopropanedioate;hydrochloride
SMILES CCOC(=O)C(C(=O)OCC)N.Cl

Specifications

Melting Point ∼165°C (decomposition)
Assay Percent Range 98%
Quantity 250 g
Beilstein 3568037
Solubility Information Soluble in water.
Formula Weight 211.65
Percent Purity 98%
Chemical Name or Material Diethyl aminomalonate hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-556-8
TSCA No
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only