missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Allylmalonate 98.0+%, TCI America™

Catalog No. p-7116776
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
A023125G 25 g
A0231500G 500 g
2 options

Catalog No. A023125G

Supplier: TCI America A023125G

Only null left

Chemical Identifiers

CAS 2049-80-1
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
MDL Number MFCD00009155
InChI Key GDWAYKGILJJNBB-UHFFFAOYSA-N
Synonym diethyl allylmalonate, ethyl allylmalonate, diethyl 2-allylmalonate, allylmalonic acid diethyl ester, diethyl prop-2-enylpropanedioate, diethyl 2-prop-2-enyl malonate, malonic acid, allyl-, diethyl ester, propanedioic acid, 2-propenyl-, diethyl ester, diethyl 2-2-propenyl-1,3-propanedioate, allyl malonic acid
PubChem CID 74900
IUPAC Name diethyl 2-prop-2-enylpropanedioate
SMILES CCOC(=O)C(CC=C)C(=O)OCC

Specifications

Color Colorless
Boiling Point 223°C
Quantity 25 g
Formula Weight 200.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Allylmalonate