missing translation for 'onlineSavingsMsg'
Learn More

Diethyl allylmalonate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 113965000

Quantity 500g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC113965000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2049-80-1
200.23
GDWAYKGILJJNBB-UHFFFAOYSA-N
74900
CCOC(=O)C(CC=C)C(=O)OCC
C10H16O4
MFCD00009155
diethyl allylmalonate, ethyl allylmalonate, diethyl 2-allylmalonate, allylmalonic acid diethyl ester, diethyl prop-2-enylpropanedioate, diethyl 2-prop-2-enyl malonate, malonic acid, allyl-, diethyl ester, propanedioic acid, 2-propenyl-, diethyl ester, diethyl 2-2-propenyl-1,3-propanedioate, allyl malonic acid
diethyl 2-prop-2-enylpropanedioate

Specifications

Diethyl allylmalonate
2049-80-1
222.0°C to 223.0°C
Authentic
Glass bottle
C2H5O2CCH(CH2CH=CH2)CO2C2H5
500g
02, 776
diethyl allylmalonate, ethyl allylmalonate, diethyl 2-allylmalonate, allylmalonic acid diethyl ester, diethyl prop-2-enylpropanedioate, diethyl 2-prop-2-enyl malonate, malonic acid, allyl-, diethyl ester, propanedioic acid, 2-propenyl-, diethyl ester, diethyl 2-2-propenyl-1,3-propanedioate, allyl malonic acid
CCOC(=O)C(CC=C)C(=O)OCC
200.23
200.23
97%
1.0100g/mL
92°C
96% min. (GC)
C10H16O4
1.4300 to 1.4320
MFCD00009155
1.01
GDWAYKGILJJNBB-UHFFFAOYSA-N
diethyl 2-prop-2-enylpropanedioate
74900
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 218-072-9

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...