Learn More
Diethyl allylmalonate, 96%
CAS: 2049-80-1 | C10H16O4 | 200.234 g/mol
Supplier: Thermo Scientific Chemicals L0228622
Description
Diethyl allylmalonate is an important intermediates in syntheses of vitamins B1 and B6, barbiturates, non-steroidal anti-inflammatory agents, other numerous pharmaceuticals, agrochemicals and flavors and fragrances compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl allylmalonate is an important intermediates in syntheses of vitamins B1 and B6, barbiturates, non-steroidal anti-inflammatory agents, other numerous pharmaceuticals, agrochemicals and flavors and fragrances compounds.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
| Diethyl allylmalonate | |
| 1.016 | |
| 92°C (197°F) | |
| 1.431 | |
| 100 g | |
| diethyl allylmalonate, ethyl allylmalonate, diethyl 2-allylmalonate, allylmalonic acid diethyl ester, diethyl prop-2-enylpropanedioate, diethyl 2-prop-2-enyl malonate, malonic acid, allyl-, diethyl ester, propanedioic acid, 2-propenyl-, diethyl ester, diethyl 2-2-propenyl-1,3-propanedioate, allyl malonic acid | |
| GDWAYKGILJJNBB-UHFFFAOYSA-N | |
| diethyl 2-prop-2-enylpropanedioate | |
| 74900 | |
| 97% |
| 2049-80-1 | |
| 222°C to 223°C | |
| C10H16O4 | |
| MFCD00009155 | |
| 1709466 | |
| Not miscible or difficult to mix with water. | |
| CCOC(=O)C(CC=C)C(=O)OCC | |
| 200.234 | |
| 200.23 |
Chemical Identifiers
| 2049-80-1 | |
| 200.234 | |
| GDWAYKGILJJNBB-UHFFFAOYSA-N | |
| 74900 | |
| CCOC(=O)C(CC=C)C(=O)OCC |
| C10H16O4 | |
| MFCD00009155 | |
| diethyl allylmalonate, ethyl allylmalonate, diethyl 2-allylmalonate, allylmalonic acid diethyl ester, diethyl prop-2-enylpropanedioate, diethyl 2-prop-2-enyl malonate, malonic acid, allyl-, diethyl ester, propanedioic acid, 2-propenyl-, diethyl ester, diethyl 2-2-propenyl-1,3-propanedioate, allyl malonic acid | |
| diethyl 2-prop-2-enylpropanedioate |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 218-072-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only