missing translation for 'onlineSavingsMsg'
Learn More

Diethyl allylmalonate, 96%

Catalog No. AAL0228614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0228614 25 g
AAL0228622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0228614 Supplier Thermo Scientific Chemicals Supplier No. L0228614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2049-80-1 | C10H16O4 | 200.234 g/mol

Diethyl allylmalonate is an important intermediates in syntheses of vitamins B1 and B6, barbiturates, non-steroidal anti-inflammatory agents, other numerous pharmaceuticals, agrochemicals and flavors and fragrances compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl allylmalonate is an important intermediates in syntheses of vitamins B1 and B6, barbiturates, non-steroidal anti-inflammatory agents, other numerous pharmaceuticals, agrochemicals and flavors and fragrances compounds.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2049-80-1
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
MDL Number MFCD00009155
InChI Key GDWAYKGILJJNBB-UHFFFAOYSA-N
Synonym diethyl allylmalonate, ethyl allylmalonate, diethyl 2-allylmalonate, allylmalonic acid diethyl ester, diethyl prop-2-enylpropanedioate, diethyl 2-prop-2-enyl malonate, malonic acid, allyl-, diethyl ester, propanedioic acid, 2-propenyl-, diethyl ester, diethyl 2-2-propenyl-1,3-propanedioate, allyl malonic acid
PubChem CID 74900
IUPAC Name diethyl 2-prop-2-enylpropanedioate
SMILES CCOC(=O)C(CC=C)C(=O)OCC

Specifications

Density 1.016
Boiling Point 222°C to 223°C
Flash Point 92°C (197°F)
Refractive Index 1.431
Quantity 25 g
Beilstein 1709466
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 200.23
Percent Purity 97%
Chemical Name or Material Diethyl allylmalonate
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 218-072-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only