Learn More
Diethyl allyl(3-phenyl-2-propynyl)malonate, 96%, Thermo Scientific Chemicals
$127.99 - $127.99
Chemical Identifiers
| CAS | 143633-91-4 |
|---|---|
| Molecular Formula | C19H22O4 |
| Molecular Weight (g/mol) | 314.381 |
| MDL Number | MFCD09953493 |
| InChI Key | ZSPBIVGRCZUELN-UHFFFAOYSA-N |
| Synonym | diethyl allyl 3-phenyl-2-propynyl malonate, diethyl 2-allyl-2-3-phenylprop-2-yn-1-yl malonate, propanedioic acid,2-3-phenyl-2-propyn-1-yl-2-2-propen-1-yl-, 1,3-diethyl ester, 1,3-diethyl 2-3-phenylprop-2-yn-1-yl-2-prop-2-en-1-yl propanedioate, acmc-20ed36, 3-phenylpropargylallylmalonic acid diethyl ester, allyl 3-phenyl-2-propynyl malonic acid diethyl ester, diethyl 1-phenyl-6-heptene-1-yne-4,4-dicarboxylate, diethyl 3-phenylprop-2-yn-1-yl prop-2-en-1-yl propanedioate |
| PubChem CID | 10086992 |
| IUPAC Name | diethyl 2-(3-phenylprop-2-ynyl)-2-prop-2-enylpropanedioate |
| SMILES | CCOC(=O)C(CC=C)(CC#CC1=CC=CC=C1)C(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2770203
|
Thermo Scientific Chemicals
H2770203 |
1 g |
Each for $127.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 143633-91-4 | |
| 314.381 | |
| ZSPBIVGRCZUELN-UHFFFAOYSA-N | |
| 10086992 | |
| CCOC(=O)C(CC=C)(CC#CC1=CC=CC=C1)C(=O)OCC |
| C19H22O4 | |
| MFCD09953493 | |
| diethyl allyl 3-phenyl-2-propynyl malonate, diethyl 2-allyl-2-3-phenylprop-2-yn-1-yl malonate, propanedioic acid,2-3-phenyl-2-propyn-1-yl-2-2-propen-1-yl-, 1,3-diethyl ester, 1,3-diethyl 2-3-phenylprop-2-yn-1-yl-2-prop-2-en-1-yl propanedioate, acmc-20ed36, 3-phenylpropargylallylmalonic acid diethyl ester, allyl 3-phenyl-2-propynyl malonic acid diethyl ester, diethyl 1-phenyl-6-heptene-1-yne-4,4-dicarboxylate, diethyl 3-phenylprop-2-yn-1-yl prop-2-en-1-yl propanedioate | |
| diethyl 2-(3-phenylprop-2-ynyl)-2-prop-2-enylpropanedioate |
Specifications
| 143633-91-4 | |
| 1.514 | |
| 1 g | |
| ZSPBIVGRCZUELN-UHFFFAOYSA-N | |
| diethyl 2-(3-phenylprop-2-ynyl)-2-prop-2-enylpropanedioate | |
| 10086992 | |
| 96% |
| C19H22O4 | |
| MFCD09953493 | |
| diethyl allyl 3-phenyl-2-propynyl malonate, diethyl 2-allyl-2-3-phenylprop-2-yn-1-yl malonate, propanedioic acid,2-3-phenyl-2-propyn-1-yl-2-2-propen-1-yl-, 1,3-diethyl ester, 1,3-diethyl 2-3-phenylprop-2-yn-1-yl-2-prop-2-en-1-yl propanedioate, acmc-20ed36, 3-phenylpropargylallylmalonic acid diethyl ester, allyl 3-phenyl-2-propynyl malonic acid diethyl ester, diethyl 1-phenyl-6-heptene-1-yne-4,4-dicarboxylate, diethyl 3-phenylprop-2-yn-1-yl prop-2-en-1-yl propanedioate | |
| CCOC(=O)C(CC=C)(CC#CC1=CC=CC=C1)C(=O)OCC | |
| 314.381 | |
| 314.38 | |
| Diethyl allyl(3-phenyl-2-propynyl)malonate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only