missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Adipate 99.0+%, TCI America™

Catalog No. A016225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A016225ML 25 mL
A0162500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A016225ML Supplier TCI America Supplier No. A016225ML
Only null left

Chemical Identifiers

CAS 141-28-6
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00009215
InChI Key VIZORQUEIQEFRT-UHFFFAOYSA-N
Synonym diethyl adipate, ethyl adipate, 1,6-diethyl hexanedioate, hexanedioic acid, diethyl ester, adipic acid, diethyl ester, adipic acid diethyl ester, hexanedioic acid, diethyl, diethylhexanedioic acid, diethylester kyseliny adipove, ethyl delta-carboethoxyvalerate
PubChem CID 8844
ChEBI CHEBI:34697
IUPAC Name diethyl hexanedioate
SMILES CCOC(=O)CCCCC(=O)OCC

Specifications

Melting Point -20°C
Color Colorless
Boiling Point 251°C
Quantity 25 mL
Formula Weight 202.25
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Adipate
EINECSNumber (2)-0861&(2)-0879
RTECSNumber AV1100000
TSCA Yes