missing translation for 'onlineSavingsMsg'
Learn More

Diethyl adipate, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 407175000

Quantity 500g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC407175000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

141-28-6
202.25
VIZORQUEIQEFRT-UHFFFAOYSA-N
8844
diethyl hexanedioate
C10H18O4
MFCD00009215
diethyl adipate, ethyl adipate, 1,6-diethyl hexanedioate, hexanedioic acid, diethyl ester, adipic acid, diethyl ester, adipic acid diethyl ester, hexanedioic acid, diethyl, diethylhexanedioic acid, diethylester kyseliny adipove, ethyl delta-carboethoxyvalerate
CHEBI:34697
CCOC(=O)CCCCC(=O)OCC

Specifications

Diethyl adipate
1.0000g/mL
>110°C
99%
C10H18O4
CH3CH2OCO(CH2)4COOCH3CH2
MFCD00009215
1
VIZORQUEIQEFRT-UHFFFAOYSA-N
diethyl hexanedioate
8844
202.25
141-28-6
132.0°C (15.0 mmHg)
Authentic
Glass bottle
1.4260 to 1.4280
500g
02, 652
diethyl adipate, ethyl adipate, 1,6-diethyl hexanedioate, hexanedioic acid, diethyl ester, adipic acid, diethyl ester, adipic acid diethyl ester, hexanedioic acid, diethyl, diethylhexanedioic acid, diethylester kyseliny adipove, ethyl delta-carboethoxyvalerate
CCOC(=O)CCCCC(=O)OCC
202.25
CHEBI:34697
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-477-

SDS
Documents

Documents

Promotions

Promotions