missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl adipate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 407171000
| Quantity | 100g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C10H18O4 | |
| MFCD00009215 | |
| diethyl adipate, ethyl adipate, 1,6-diethyl hexanedioate, hexanedioic acid, diethyl ester, adipic acid, diethyl ester, adipic acid diethyl ester, hexanedioic acid, diethyl, diethylhexanedioic acid, diethylester kyseliny adipove, ethyl delta-carboethoxyvalerate | |
| CHEBI:34697 | |
| CCOC(=O)CCCCC(=O)OCC |
Specifications
| Diethyl adipate | |
| 1.0000g/mL | |
| >110°C | |
| 99% | |
| C10H18O4 | |
| CH3CH2OCO(CH2)4COOCH3CH2 | |
| MFCD00009215 | |
| 1 | |
| VIZORQUEIQEFRT-UHFFFAOYSA-N | |
| diethyl hexanedioate | |
| 8844 | |
| 202.25 |
| 141-28-6 | |
| 132.0°C (15.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4260 to 1.4280 | |
| 100g | |
| 02, 652 | |
| diethyl adipate, ethyl adipate, 1,6-diethyl hexanedioate, hexanedioic acid, diethyl ester, adipic acid, diethyl ester, adipic acid diethyl ester, hexanedioic acid, diethyl, diethylhexanedioic acid, diethylester kyseliny adipove, ethyl delta-carboethoxyvalerate | |
| CCOC(=O)CCCCC(=O)OCC | |
| 202.25 | |
| CHEBI:34697 | |
| 99% |
Safety and Handling
EINECSNumber : 205-477-