missing translation for 'onlineSavingsMsg'
Learn More

Diethyl adipate, 99%

Catalog No. AAA1147622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1147622 100 g
AAA1147636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1147622 Supplier Thermo Scientific Chemicals Supplier No. A1147622
Only null left

CAS: 141-28-6 | C10H18O4 | 202.25 g/mol

Poly(vinylidene fluoride) (PVF2) gels in diethyl adipate (DEA) with fibrillar morphology is presented. Gelation process obeys the three-dimensional percolation mechanism. Combustion and emission characteristics of a direct-injection diesel engine is fueled with diesel-diethyl adipate blends.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Poly(vinylidene fluoride) (PVF2) gels in diethyl adipate (DEA) with fibrillar morphology is presented. Gelation process obeys the three-dimensional percolation mechanism. Combustion and emission characteristics of a direct-injection diesel engine is fueled with diesel-diethyl adipate blends.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 141-28-6
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00009215
InChI Key VIZORQUEIQEFRT-UHFFFAOYSA-N
Synonym diethyl adipate, ethyl adipate, 1,6-diethyl hexanedioate, hexanedioic acid, diethyl ester, adipic acid, diethyl ester, adipic acid diethyl ester, hexanedioic acid, diethyl, diethylhexanedioic acid, diethylester kyseliny adipove, ethyl delta-carboethoxyvalerate
PubChem CID 8844
ChEBI CHEBI:34697
IUPAC Name diethyl hexanedioate
SMILES CCOC(=O)CCCCC(=O)OCC

Specifications

Melting Point -20°C
Density 1.008
Boiling Point 250°C to 252°C
Flash Point >110°C (230°F)
Refractive Index 1.4275
Linear Formula CH3CH2O2C(CH2)4CO2CH2CH3
Quantity 100 g
Beilstein 1780035
Merck Index 14,162
Solubility Information Insoluble in water.
Formula Weight 202.25
Percent Purity 99%
Chemical Name or Material Diethyl adipate
Show More Show Less
EINECSNumber 205-477-0
RTECSNumber AV1100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only