missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Acetonylmalonate 95.0+%, TCI America™

Catalog No. A15035G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15035G 5 g
A15031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15035G Supplier TCI America Supplier No. A15035G

Chemical Identifiers

CAS 23193-18-2
Molecular Formula C10H16O5
Molecular Weight (g/mol) 216.233
MDL Number MFCD00177949
InChI Key KGZCSZOGMMZHKB-UHFFFAOYSA-N
Synonym Acetonylmalonic Acid Diethyl Ester, Diethyl 2-Oxopropylmalonate, 2-Oxopropylmalonic Acid Diethyl Ester
PubChem CID 12000252
IUPAC Name diethyl 2-(2-oxopropyl)propanedioate
SMILES CCOC(=O)C(CC(=O)C)C(=O)OCC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 216.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Acetonylmalonate