missing translation for 'onlineSavingsMsg'
Learn More

Diethyl acetone-1,3-dicarboxylate, 96%

Catalog No. AAA1093330
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1093330 250 g
AAA1093318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1093330 Supplier Thermo Scientific Chemicals Supplier No. A1093330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 105-50-0 | C9H14O5 | 202.21 g/mol

Synthesis of 1-substituted 4-ethoxycarbonyl-5- (ethoxycarbonylmethyl) pyrazoles Diethyl acetone-1, 3-dicarboxylate reacts with N, N-dimethylforma- mide dimethyl acetal (DMFDMA) in ethanol at room temperature. Unexpected product dichotomy is produced in the Biginelli-like condensation of 2-hydroxybenzaldehyde with urea or thiourea and dimethyl or diethyl acetone-1,3-dicarboxylate, respectively, as active methylene components. Ethyl 2-amino-4-(2-ethoxy-2-oxoethyl)thiazole-5-carboxylate (2a), prepared from diethylacetone-1,3-dicarboxylate, sulfuryl chloride and thiourea. Synthesis of diethyl 2,2-diethyl-3,5-dioxopimelate by the reaction of ethyl 3-chloro-3-oxo-2,2-dimethylpropionate with diethyl acetone-1,3-dicarboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Synthesis of 1-substituted 4-ethoxycarbonyl-5- (ethoxycarbonylmethyl) pyrazoles Diethyl acetone-1, 3-dicarboxylate reacts with N, N-dimethylforma- mide dimethyl acetal (DMFDMA) in ethanol at room temperature. Unexpected product dichotomy is produced in the Biginelli-like condensation of 2-hydroxybenzaldehyde with urea or thiourea and dimethyl or diethyl acetone-1,3-dicarboxylate, respectively, as active methylene components. Ethyl 2-amino-4-(2-ethoxy-2-oxoethyl)thiazole-5-carboxylate (2a), prepared from diethylacetone-1,3-dicarboxylate, sulfuryl chloride and thiourea. Synthesis of diethyl 2,2-diethyl-3,5-dioxopimelate by the reaction of ethyl 3-chloro-3-oxo-2,2-dimethylpropionate with diethyl acetone-1,3-dicarboxylate.

Solubility
Insoluble in water and soluble in alcohol, ester, benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 105-50-0
Molecular Formula C9H14O5
Molecular Weight (g/mol) 202.21
MDL Number MFCD00009200
InChI Key ZSANYRMTSBBUCA-UHFFFAOYSA-N
Synonym diethyl 1,3-acetonedicarboxylate, diethyl 3-oxoglutarate, 1,5-diethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, diethyl ester, ethyl acetonedicarboxylate, ethyl 3-oxoglutarate, diethyl acetone-1,3-dicarboxylate, diethyl acetone dicarboxylate, 3-oxopentanedioic acid diethyl ester, acetonedicarboxylic acid diethyl ester
PubChem CID 66045
IUPAC Name diethyl 3-oxopentanedioate
SMILES CCOC(=O)CC(=O)CC(=O)OCC

Specifications

Density 1.113
Boiling Point 134°C to 135°C (10 mmHg)
Flash Point 86°C (186°F)
Refractive Index 1.44
Quantity 250 g
Beilstein 640146
Solubility Information Insoluble in water and soluble in alcohol,ester,benzene.
Formula Weight 202.21
Percent Purity 96%
Chemical Name or Material Diethyl acetone-1,3-dicarboxylate
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 203-302-2
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only