missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Acetamidomalonate 98.0+%, TCI America™
$48.13 - $306.10
Chemical Identifiers
| CAS | 1068-90-2 |
|---|---|
| Molecular Formula | C9H15NO5 |
| Molecular Weight (g/mol) | 217.221 |
| MDL Number | MFCD00009146 |
| InChI Key | ISOLMABRZPQKOV-UHFFFAOYSA-N |
| Synonym | diethyl acetamidomalonate, diethyl 2-acetamidomalonate, diethyl acetaminomalonate, diethyl acetylaminomalonate, acetamidomalonic acid diethyl ester, 2-acetylaminomalonic acid diethyl ester, 1,3-diethyl 2-acetamidopropanedioate, diethyl acetylamino malonate, propanedioic acid, acetylamino-, diethyl ester, acetylamino propanedioic acid diethyl ester |
| PubChem CID | 14041 |
| IUPAC Name | diethyl 2-acetamidopropanedioate |
| SMILES | CCOC(=O)C(C(=O)OCC)NC(=O)C |
Chemical Identifiers
| 1068-90-2 | |
| 217.221 | |
| ISOLMABRZPQKOV-UHFFFAOYSA-N | |
| 14041 | |
| CCOC(=O)C(C(=O)OCC)NC(=O)C |
| C9H15NO5 | |
| MFCD00009146 | |
| diethyl acetamidomalonate, diethyl 2-acetamidomalonate, diethyl acetaminomalonate, diethyl acetylaminomalonate, acetamidomalonic acid diethyl ester, 2-acetylaminomalonic acid diethyl ester, 1,3-diethyl 2-acetamidopropanedioate, diethyl acetylamino malonate, propanedioic acid, acetylamino-, diethyl ester, acetylamino propanedioic acid diethyl ester | |
| diethyl 2-acetamidopropanedioate |
Specifications
| 1068-90-2 | |
| White | |
| MFCD00009146 | |
| diethyl acetamidomalonate, diethyl 2-acetamidomalonate, diethyl acetaminomalonate, diethyl acetylaminomalonate, acetamidomalonic acid diethyl ester, 2-acetylaminomalonic acid diethyl ester, 1,3-diethyl 2-acetamidopropanedioate, diethyl acetylamino malonate, propanedioic acid, acetylamino-, diethyl ester, acetylamino propanedioic acid diethyl ester | |
| CCOC(=O)C(C(=O)OCC)NC(=O)C | |
| 217.221 | |
| 217.22 | |
| Crystalline Powder |
| 96°C | |
| C9H15NO5 | |
| 25 g | |
| ISOLMABRZPQKOV-UHFFFAOYSA-N | |
| diethyl 2-acetamidopropanedioate | |
| 14041 | |
| ≥98.0% (GC) | |
| Diethyl Acetamidomalonate |
Safety and Handling
EINECSNumber : (9)-1636
RTECSNumber : OO0360000
TSCA : Yes