missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Acetamidomalonate 98.0+%, TCI America™

Catalog No. A001525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A001525G 25 g
A0015100G 100 g
A0015500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A001525G Supplier TCI America Supplier No. A001525G

Chemical Identifiers

CAS 1068-90-2
Molecular Formula C9H15NO5
Molecular Weight (g/mol) 217.221
MDL Number MFCD00009146
InChI Key ISOLMABRZPQKOV-UHFFFAOYSA-N
Synonym diethyl acetamidomalonate, diethyl 2-acetamidomalonate, diethyl acetaminomalonate, diethyl acetylaminomalonate, acetamidomalonic acid diethyl ester, 2-acetylaminomalonic acid diethyl ester, 1,3-diethyl 2-acetamidopropanedioate, diethyl acetylamino malonate, propanedioic acid, acetylamino-, diethyl ester, acetylamino propanedioic acid diethyl ester
PubChem CID 14041
IUPAC Name diethyl 2-acetamidopropanedioate
SMILES CCOC(=O)C(C(=O)OCC)NC(=O)C

Specifications

Melting Point 96°C
Color White
Quantity 25 g
Formula Weight 217.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl Acetamidomalonate
EINECSNumber (9)-1636
RTECSNumber OO0360000
TSCA Yes