missing translation for 'onlineSavingsMsg'
Learn More

Diethyl acetamidomalonate, 98+%

Catalog No. p-7021637
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1404814 25 g
AAA1404822 100 g
AAA1404836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1404814 Supplier Thermo Scientific Chemicals Supplier No. A1404814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1068-90-2 | C9H15NO5 | 217.221 g/mol

Diethyl 2-Acetamidomalonate, is a versatile building block used for the synthesis of various pharmaceutical and biologically active compounds. It is an intermediate for the preparation of Novobiocin analogues as potential heat shock protein 90 inhibitors. It is also used as a important intermediates in syntheses of vitamins B1 and B6, barbiturates, non-steroidal anti-inflammatory agents, other numerous pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 2-Acetamidomalonate, is a versatile building block used for the synthesis of various pharmaceutical and biologically active compounds. It is an intermediate for the preparation of Novobiocin analogues as potential heat shock protein 90 inhibitors. It is also used as a important intermediates in syntheses of vitamins B1 and B6, barbiturates, non-steroidal anti-inflammatory agents, other numerous pharmaceuticals.

Solubility
Soluble in chloroform and methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.

Chemical Identifiers

CAS 1068-90-2
Molecular Formula C9H15NO5
Molecular Weight (g/mol) 217.221
MDL Number MFCD00009146
InChI Key ISOLMABRZPQKOV-UHFFFAOYSA-N
Synonym diethyl acetamidomalonate, diethyl 2-acetamidomalonate, diethyl acetaminomalonate, diethyl acetylaminomalonate, acetamidomalonic acid diethyl ester, 2-acetylaminomalonic acid diethyl ester, 1,3-diethyl 2-acetamidopropanedioate, diethyl acetylamino malonate, propanedioic acid, acetylamino-, diethyl ester, acetylamino propanedioic acid diethyl ester
PubChem CID 14041
IUPAC Name diethyl 2-acetamidopropanedioate
SMILES CCOC(=O)C(C(=O)OCC)NC(=O)C

Specifications

Melting Point 96°C to 99°C
Boiling Point 185°C (20 mmHg)
Quantity 25 g
Beilstein 783883
Solubility Information Soluble in chloroform and methanol. Slightly soluble in water.
Formula Weight 217.22
Percent Purity ≥98%
Chemical Name or Material Diethyl acetamidomalonate
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 213-952-9
RTECSNumber OO0360000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only