missing translation for 'onlineSavingsMsg'
Learn More

Diethyl acetamidomalonate, 98+%

CAS: 1068-90-2 | C9H15NO5 | 217.221 g/mol

Supplier:  Thermo Scientific Chemicals A1404822

 View more versions of this product

Catalog No. AAA1404822


Only null left
Explore available promotions

Description

Description

Diethyl 2-Acetamidomalonate, is a versatile building block used for the synthesis of various pharmaceutical and biologically active compounds. It is an intermediate for the preparation of Novobiocin analogues as potential heat shock protein 90 inhibitors. It is also used as a important intermediates in syntheses of vitamins B1 and B6, barbiturates, non-steroidal anti-inflammatory agents, other numerous pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 2-Acetamidomalonate, is a versatile building block used for the synthesis of various pharmaceutical and biologically active compounds. It is an intermediate for the preparation of Novobiocin analogues as potential heat shock protein 90 inhibitors. It is also used as a important intermediates in syntheses of vitamins B1 and B6, barbiturates, non-steroidal anti-inflammatory agents, other numerous pharmaceuticals.

Solubility
Soluble in chloroform and methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.
Specifications

Specifications

Diethyl acetamidomalonate
96°C to 99°C
C9H15NO5
100 g
diethyl acetamidomalonate, diethyl 2-acetamidomalonate, diethyl acetaminomalonate, diethyl acetylaminomalonate, acetamidomalonic acid diethyl ester, 2-acetylaminomalonic acid diethyl ester, 1,3-diethyl 2-acetamidopropanedioate, diethyl acetylamino malonate, propanedioic acid, acetylamino-, diethyl ester, acetylamino propanedioic acid diethyl ester
ISOLMABRZPQKOV-UHFFFAOYSA-N
diethyl 2-acetamidopropanedioate
14041
≥98%
1068-90-2
185°C (20 mmHg)
MFCD00009146
783883
Soluble in chloroform and methanol. Slightly soluble in water.
CCOC(=O)C(C(=O)OCC)NC(=O)C
217.221
217.22

Chemical Identifiers

1068-90-2
217.221
ISOLMABRZPQKOV-UHFFFAOYSA-N
14041
CCOC(=O)C(C(=O)OCC)NC(=O)C
C9H15NO5
MFCD00009146
diethyl acetamidomalonate, diethyl 2-acetamidomalonate, diethyl acetaminomalonate, diethyl acetylaminomalonate, acetamidomalonic acid diethyl ester, 2-acetylaminomalonic acid diethyl ester, 1,3-diethyl 2-acetamidopropanedioate, diethyl acetylamino malonate, propanedioic acid, acetylamino-, diethyl ester, acetylamino propanedioic acid diethyl ester
diethyl 2-acetamidopropanedioate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 213-952-9

RTECSNumber : OO0360000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only