missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 5-Amino-3-methyl-2,4-thiophenedicarboxylate 98.0+%, TCI America™

Catalog No. A13545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13545G 5 g
A135425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13545G Supplier TCI America Supplier No. A13545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4815-30-9
Molecular Formula C11H15NO4S
Molecular Weight (g/mol) 257.30
MDL Number MFCD00005450
InChI Key DGVXLHAJVRRLGV-UHFFFAOYSA-N
Synonym diethyl 5-amino-3-methyl-2,4-thiophenedicarboxylate, 2,4-diethyl 5-amino-3-methylthiophene-2,4-dicarboxylate, diethyl 2-amino-4-methylthiophene-3,5-dicarboxylate, diethyl 5-amino-3-methyl-thiophene-2,4-dicarboxylate, diethyl5-amino-3-methylthiophene-2,4-dicarboxylate, 2,4-thiophenedicarboxylic acid, 5-amino-3-methyl-, diethyl ester, 2-amino-3,5-bis ethoxycarbonyl-4-methylthiophene, 5-amino-3-methyl-thiophene-2,4-dicarboxylic acid diethyl ester, 2-amino-4-methyl-3,5-bis ethoxycarbonyl thiophene, diethyl 5-amino-3-methyl-2,4-thiophene dicarboxylate
PubChem CID 78537
IUPAC Name 2,4-diethyl 5-amino-3-methylthiophene-2,4-dicarboxylate
SMILES CCOC(=O)C1=C(C)C(C(=O)OCC)=C(N)S1

Specifications

Melting Point 108°C
Quantity 5 g
Formula Weight 257.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl 5-Amino-3-methyl-2,4-thiophenedicarboxylate