missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (4-nitrobenzyl)phosphonate, 98%

Catalog No. p-109948
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC332290050 5 g Glass bottle
AC332290250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC332290050 Supplier Thermo Scientific Chemicals Supplier No. 332290050

CAS: 2609-49-6 | C11H16NO5P | 273.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 2609-49-6
Molecular Formula C11H16NO5P
Molecular Weight (g/mol) 273.22
MDL Number MFCD00047747
InChI Key FORMFFDDQMCTCT-UHFFFAOYSA-N
PubChem CID 256065
IUPAC Name 1-(diethoxyphosphorylmethyl)-4-nitrobenzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)[N+](=O)[O-])OCC

Specifications

Color Orange-Yellow to Red
Density 1.2400g/mL
Boiling Point 151°C (0.1 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5220 to 1.526
Quantity 5 g
Beilstein 16, IV, 1096
Specific Gravity 1.24
Synonym diethyl 4-nitrobenzylphosphonate, diethyl 4-nitrobenzyl phosphonate, diethyl 4-nitrophenyl methylphosphonate, 1-diethoxyphosphorylmethyl-4-nitro-benzene, 1-diethoxyphosphorylmethyl-4-nitrobenzene, phosphonic acid, 4-nitrophenyl methyl-, diethyl ester, phosphonic acid, p-4-nitrophenyl methyl-, diethyl ester, zlchem 660, diethyl4-nitrobenzylphosphonate
Formula Weight 273.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diethyl (4-nitrobenzyl)phosphonate
Show More Show Less

RUO – Research Use Only