missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (4-Methylbenzyl)phosphonate 98.0+%, TCI America™

Catalog No. p-7127673
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33365G 5 g
D333625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33365G Supplier TCI America Supplier No. D33365G
Only null left

Chemical Identifiers

CAS 3762-25-2
Molecular Formula C12H19O3P
Molecular Weight (g/mol) 242.255
MDL Number MFCD00041385
InChI Key QKGBKPZAXXBLJE-UHFFFAOYSA-N
Synonym diethyl 4-methylbenzylphosphonate, diethyl 4-methylbenzyl phosphonate, 4-methylbenzyl phosphonic acid diethyl ester, diethyl 4-methylphenyl methylphosphonate, diethoxy 4-methylphenyl methyl phosphino-1-one, acmc-209iuf, diethyl-4-methylbenzylphosphonate, 1-diethoxyphosphorylmethyl-4-methylbenzene, phosphonic acid, 4-methylphenyl methyl-, diethyl ester
PubChem CID 3050
IUPAC Name 1-(diethoxyphosphorylmethyl)-4-methylbenzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)C)OCC

Specifications

Color Yellow
Boiling Point 160°C
Quantity 5 g
Formula Weight 242.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (4-Methylbenzyl)phosphonate