missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 4-methoxybenzylphosphonate, 98+%

Catalog No. AAL0522609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0522609 10 g
AAL0522618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0522609 Supplier Thermo Scientific Chemicals Supplier No. L0522609

CAS: 1145-93-3 | C12H19O4P | 258.254 g/mol

Diethyl 4-methoxybenzylphosphonate is used in synthesis of C-aryl glycosides. It acts as a reactant for synthesis of γ -Monofluorinated goniothalamin analogs via ring-opening hydrofluorination, stilbenoid derivatives with neuroprotective activity, resveratrol-chroman hybrids with antioxidant activity, Oligostilbenoids using regioselective cyclodehydration and arylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 4-methoxybenzylphosphonate is used in synthesis of C-aryl glycosides. It acts as a reactant for synthesis of γ -Monofluorinated goniothalamin analogs via ring-opening hydrofluorination, stilbenoid derivatives with neuroprotective activity, resveratrol-chroman hybrids with antioxidant activity, Oligostilbenoids using regioselective cyclodehydration and arylation.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.

Chemical Identifiers

CAS 1145-93-3
Molecular Formula C12H19O4P
Molecular Weight (g/mol) 258.254
MDL Number MFCD00015662
InChI Key NKARVHNAACNYGE-UHFFFAOYSA-N
Synonym diethyl 4-methoxybenzylphosphonate, diethyl 4-methoxybenzyl phosphonate, 1-diethoxyphosphorylmethyl-4-methoxybenzene, diethyl 4-methoxyphenyl methylphosphonate, 1-diethoxyphosphorylmethyl-4-methoxy-benzene, phosphonic acid, 4-methoxyphenyl methyl-, diethyl ester, acmc-1bsuk, 4-diethylphosphonomethyl anisole, diethyl-4-methoxybenzyl phosphonate, diethyl 4-methoxybenzyl-phosphonate
PubChem CID 4063826
IUPAC Name 1-(diethoxyphosphorylmethyl)-4-methoxybenzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)OC)OCC

Specifications

Density 1.5
Boiling Point 136°C to 138°C (0.7 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5045
Quantity 10 g
Beilstein 2125148
Solubility Information Not miscible in water.
Formula Weight 258.25
Percent Purity ≥98%
Chemical Name or Material Diethyl 4-methoxybenzylphosphonate
EINECSNumber 214-548-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only