missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (4-Methoxybenzyl)phosphonate 95.0+%, TCI America™

Catalog No. D40005G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40005G 5 g
D400025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40005G Supplier TCI America Supplier No. D40005G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1145-93-3
Molecular Formula C12H19O4P
Molecular Weight (g/mol) 258.254
MDL Number MFCD00015662
InChI Key NKARVHNAACNYGE-UHFFFAOYSA-N
Synonym diethyl 4-methoxybenzylphosphonate, diethyl 4-methoxybenzyl phosphonate, 1-diethoxyphosphorylmethyl-4-methoxybenzene, diethyl 4-methoxyphenyl methylphosphonate, 1-diethoxyphosphorylmethyl-4-methoxy-benzene, phosphonic acid, 4-methoxyphenyl methyl-, diethyl ester, acmc-1bsuk, 4-diethylphosphonomethyl anisole, diethyl-4-methoxybenzyl phosphonate, diethyl 4-methoxybenzyl-phosphonate
PubChem CID 4063826
IUPAC Name 1-(diethoxyphosphorylmethyl)-4-methoxybenzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)OC)OCC

Specifications

Color Yellow
Boiling Point 151°C
Quantity 5 g
Formula Weight 258.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (4-Methoxybenzyl)phosphonate
EINECSNumber (3)-3918
TSCA No