missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (4-Chlorobenzyl)phosphonate 97.0+%, TCI America™

Catalog No. p-7127672
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33355G 5 g
D333525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33355G Supplier TCI America Supplier No. D33355G
Only null left

Chemical Identifiers

CAS 39225-17-7
Molecular Formula C11H16ClO3P
Molecular Weight (g/mol) 262.67
InChI Key BVQPVBZRJSFOEZ-UHFFFAOYSA-N
Synonym diethyl 4-chlorobenzylphosphonate, diethyl 4-chlorobenzyl phosphonate, diethyl 4-chlorophenyl methylphosphonate, 1-chloro-4-diethoxyphosphorylmethyl benzene, phosphonic acid, 4-chlorophenyl methyl-, diethyl ester, phosphonic acid, p-4-chlorophenyl methyl-, diethyl ester, acmc-1aifv, diethyl 4-chlorobenzylposphonate, diethyl 4-chlorobenzylphosphate
PubChem CID 99240
IUPAC Name 1-chloro-4-(diethoxyphosphorylmethyl)benzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)Cl)OCC

Specifications

Color Colorless
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 180°C
Formula Weight 262.67
Percent Purity ≥97.0% (GC)
Chemical Name or Material Diethyl (4-Chlorobenzyl)phosphonate
EINECSNumber (3)-3977
TSCA No