Learn More
Diethyl 4-aminobenzylphosphonate, 99%
CAS: 20074-79-7 | C11H18NO3P | 243.24 g/mol
Supplier: Thermo Scientific Chemicals B2227914
Description
Diethyl 4-aminobenzylphosphonate is used as a reagent in the preparation of [4-(tetradecanoylamino)benzyl]phosphonic acid. It is also used in the production of diethyl p-benzoylaminobenzylphosphonate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl 4-aminobenzylphosphonate is used as a reagent in the preparation of [4-(tetradecanoylamino)benzyl]phosphonic acid. It is also used in the production of diethyl p-benzoylaminobenzylphosphonate.
Solubility
Soluble in methylene chloride, acetone and benzene. Insoluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
| Diethyl 4-aminobenzylphosphonate | |
| 91°C to 94°C | |
| MFCD00007913 | |
| 2941613 | |
| Soluble in methylene chloride,acetone and benzene. Insoluble in water. | |
| CCOP(=O)(CC1=CC=C(N)C=C1)OCC | |
| 243.24 | |
| 243.24 |
| 20074-79-7 | |
| C11H18NO3P | |
| 25 g | |
| diethyl 4-aminobenzylphosphonate, diethyl p-aminobenzylphosphonate, 4-diethoxyphosphorylmethyl aniline, p-aminobenzylphosphonic acid diethyl ester, diethyl p-aminobenzyl phosphonate, phosphonic acid, p-aminobenzyl-, diethyl ester, diethyl 4-aminophenyl methylphosphonate, phosphonic acid, 4-aminophenyl methyl-, diethyl ester, timtec-bb sbb000830 | |
| ZVAYUUUQOCPZCZ-UHFFFAOYSA-N | |
| 4-(diethoxyphosphorylmethyl)aniline | |
| 88358 | |
| 99% |
Chemical Identifiers
| 20074-79-7 | |
| 243.24 | |
| ZVAYUUUQOCPZCZ-UHFFFAOYSA-N | |
| 88358 |
| C11H18NO3P | |
| MFCD00007913 | |
| diethyl 4-aminobenzylphosphonate, diethyl p-aminobenzylphosphonate, 4-diethoxyphosphorylmethyl aniline, p-aminobenzylphosphonic acid diethyl ester, diethyl p-aminobenzyl phosphonate, phosphonic acid, p-aminobenzyl-, diethyl ester, diethyl 4-aminophenyl methylphosphonate, phosphonic acid, 4-aminophenyl methyl-, diethyl ester, timtec-bb sbb000830 | |
| CCOP(=O)(CC1=CC=C(N)C=C1)OCC |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 243-504-8
RTECSNumber : SZ6480000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only