missing translation for 'onlineSavingsMsg'
Learn More

Diethyl(3-pyridyl)borane 98.0+%, TCI America™

Catalog No. D26611G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26611G Supplier TCI America Supplier No. D26611G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89878-14-8
Molecular Formula C9H14BN
Molecular Weight (g/mol) 147.03
MDL Number MFCD00012348
InChI Key OJKBCQOJVMAHDX-UHFFFAOYSA-N
Synonym diethyl 3-pyridyl borane, 3-diethylboryl pyridine, diethyl 3-pyridinyl borane, pyridine, 3-diethylboryl, 3-diethylboranyl pyridine, diethyl-3-pyridylborane, dithyl 3-pyridyl borane, 3-pyridyldiethylborane, diethyl 3-pyridyl bo
PubChem CID 642851
IUPAC Name 3-(diethylboranyl)pyridine
SMILES CCB(CC)C1=CN=CC=C1

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 1 g
Formula Weight 147.03
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl(3-pyridyl)borane