missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 3-Oxopimelate 96.0+%, TCI America™
$143.54 - $417.56
Chemical Identifiers
| CAS | 40420-22-2 |
|---|---|
| Molecular Formula | C11H18O5 |
| Molecular Weight (g/mol) | 230.26 |
| MDL Number | MFCD00009214 |
| InChI Key | KAMXUPAAAGYEDK-UHFFFAOYSA-N |
| Synonym | diethyl 3-oxopimelate, diethyl3-oxopimelate, heptanedioic acid, 3-oxo-, diethyl ester, 1,7-diethyl 3-oxoheptanedioate, 3-oxopimelic acid diethyl ester, diethyl3-oxoheptanedioate, acmc-1akc9, diethyl-3-oxopimelate, diethyl 3-oxoheptane-1,7-dioate |
| PubChem CID | 575510 |
| IUPAC Name | 1,5-diethyl 2,2-dimethyl-3-oxopentanedioate |
| SMILES | CCOC(=O)CC(=O)C(C)(C)C(=O)OCC |
Chemical Identifiers
| 40420-22-2 | |
| 230.26 | |
| KAMXUPAAAGYEDK-UHFFFAOYSA-N | |
| 575510 | |
| CCOC(=O)CC(=O)C(C)(C)C(=O)OCC |
| C11H18O5 | |
| MFCD00009214 | |
| diethyl 3-oxopimelate, diethyl3-oxopimelate, heptanedioic acid, 3-oxo-, diethyl ester, 1,7-diethyl 3-oxoheptanedioate, 3-oxopimelic acid diethyl ester, diethyl3-oxoheptanedioate, acmc-1akc9, diethyl-3-oxopimelate, diethyl 3-oxoheptane-1,7-dioate | |
| 1,5-diethyl 2,2-dimethyl-3-oxopentanedioate |
Specifications
| 40420-22-2 | |
| 132°C | |
| MFCD00009214 | |
| diethyl 3-oxopimelate, diethyl3-oxopimelate, heptanedioic acid, 3-oxo-, diethyl ester, 1,7-diethyl 3-oxoheptanedioate, 3-oxopimelic acid diethyl ester, diethyl3-oxoheptanedioate, acmc-1akc9, diethyl-3-oxopimelate, diethyl 3-oxoheptane-1,7-dioate | |
| CCOC(=O)CC(=O)C(C)(C)C(=O)OCC | |
| 230.26 | |
| 230.26 | |
| Liquid |
| Yellow | |
| C11H18O5 | |
| 1 g | |
| KAMXUPAAAGYEDK-UHFFFAOYSA-N | |
| 1,5-diethyl 2,2-dimethyl-3-oxopentanedioate | |
| 575510 | |
| ≥96.0% (T) | |
| Diethyl 3-Oxopimelate |
Safety and Handling
TSCA : No