missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 3-Oxopimelate 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 132°C |
| Quantity | 5 g |
| Formula Weight | 230.26 |
| Percent Purity | ≥96.0% (T) |
| Physical Form | Liquid |
| Chemical Name or Material | Diethyl 3-Oxopimelate |
Safety and Handling
| TSCA | No |