missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 3-Hydroxyglutarate 95.0+%, TCI America™
Supplier: TCI America D319625G
Specifications
| Diethyl 3-Hydroxyglutarate | |
| Yellow | |
| C9H16O5 | |
| 3-Hydroxyglutaric Acid Diethyl Ester | |
| CCOC(=O)CC(CC(=O)OCC)O | |
| 204.222 | |
| 204.22 | |
| Liquid |
| 32328-03-3 | |
| 107°C | |
| 25 g | |
| OLLQYIBTJXUEEX-UHFFFAOYSA-N | |
| diethyl 3-hydroxypentanedioate | |
| 96259 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 32328-03-3 | |
| 204.222 | |
| 3-Hydroxyglutaric Acid Diethyl Ester | |
| diethyl 3-hydroxypentanedioate |
| C9H16O5 | |
| OLLQYIBTJXUEEX-UHFFFAOYSA-N | |
| 96259 | |
| CCOC(=O)CC(CC(=O)OCC)O |
Safety and Handling
TSCA : No