missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (3-aminopropyl)phosphonate, 95%, Thermo Scientific Chemicals

$1193.34 - $1193.34

Chemical Identifiers

CAS 4402-24-8
Molecular Formula C7H18NO3P
Molecular Weight (g/mol) 195.20
MDL Number MFCD11110596
InChI Key DQORFBNFNLHVIM-UHFFFAOYSA-N
Synonym diethyl 3-aminopropyl phosphonate, diethyl 3-aminopropylphosphonate, 3-aminopropyl phosphonic acid diethyl ester, diethyl aminopropyl phosphonate, 3-aminopropylphosphonic acid diethyl ester, 3-aminopropanephosphonic acid diethyl ester, 3-amino-propyl-phosphonic acid diethyl ester, diethyl 3-aminopropyl phosphonate 250mg, phosphonic acid, 3-aminopropyl-, diethyl ester
PubChem CID 78113
IUPAC Name 3-diethoxyphosphorylpropan-1-amine
SMILES CCOP(=O)(CCCN)OCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5187403
View Documents
Thermo Scientific Chemicals
H5187403
1 g
Each for $1,193.34
Only null left
 
Description

Description

Diethyl (3-aminopropyl)phosphonate is used in the preparation of tert-Butyl (2-((diethoxyphosphoryl)propyl)amino)-3,4-dioxocyclobut-1- en-1-yl)carbamate by reacting with tert-butyl (2-ethoxy-3,4-dioxocyclobut-1-en-1-yl)carbamate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl (3-aminopropyl)phosphonate is used in the preparation of tert-Butyl (2-((diethoxyphosphoryl)propyl)amino)-3,4-dioxocyclobut-1- en-1-yl)carbamate by reacting with tert-butyl (2-ethoxy-3,4-dioxocyclobut-1-en-1-yl)carbamate.

Notes
Air sensitive. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

4402-24-8
195.20
DQORFBNFNLHVIM-UHFFFAOYSA-N
78113
CCOP(=O)(CCCN)OCC
C7H18NO3P
MFCD11110596
diethyl 3-aminopropyl phosphonate, diethyl 3-aminopropylphosphonate, 3-aminopropyl phosphonic acid diethyl ester, diethyl aminopropyl phosphonate, 3-aminopropylphosphonic acid diethyl ester, 3-aminopropanephosphonic acid diethyl ester, 3-amino-propyl-phosphonic acid diethyl ester, diethyl 3-aminopropyl phosphonate 250mg, phosphonic acid, 3-aminopropyl-, diethyl ester
3-diethoxyphosphorylpropan-1-amine

Specifications

4402-24-8
C7H18NO3P
1 g
Air sensitive
DQORFBNFNLHVIM-UHFFFAOYSA-N
3-diethoxyphosphorylpropan-1-amine
78113
95%
1.053
MFCD11110596
UN2735
diethyl 3-aminopropyl phosphonate, diethyl 3-aminopropylphosphonate, 3-aminopropyl phosphonic acid diethyl ester, diethyl aminopropyl phosphonate, 3-aminopropylphosphonic acid diethyl ester, 3-aminopropanephosphonic acid diethyl ester, 3-amino-propyl-phosphonic acid diethyl ester, diethyl 3-aminopropyl phosphonate 250mg, phosphonic acid, 3-aminopropyl-, diethyl ester
CCOP(=O)(CCCN)OCC
195.20
195.2
Diethyl (3-aminopropyl)phosphonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 224-535-6

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only