missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-Propyl-1H-imidazole-4,5-dicarboxylate 98.0+%, TCI America™

Catalog No. D37655G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37655G 5 g
D376525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37655G Supplier TCI America Supplier No. D37655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 144689-94-1
Molecular Formula C12H18N2O4
Molecular Weight (g/mol) 254.286
MDL Number MFCD08062365
InChI Key ZQPMIDPIXSMOAD-UHFFFAOYSA-N
Synonym 2-Propyl-1H-imidazole-4,5-dicarboxylic Acid Diethyl Ester
PubChem CID 10286015
IUPAC Name diethyl 2-propyl-1H-imidazole-4,5-dicarboxylate
SMILES CCCC1=NC(=C(N1)C(=O)OCC)C(=O)OCC

Specifications

Melting Point 86°C
Color Yellow
Quantity 5 g
Formula Weight 254.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl 2-Propyl-1H-imidazole-4,5-dicarboxylate
EINECSNumber (5)-6713
TSCA No