missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (2-Oxo-2-phenylethyl)phosphonate 97.0+%, TCI America™

Catalog No. D33395G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33395G 5 g
D33391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33395G Supplier TCI America Supplier No. D33395G
Only null left

Chemical Identifiers

CAS 3453-00-7
Molecular Formula C12H17O4P
Molecular Weight (g/mol) 256.24
MDL Number MFCD00015342
InChI Key HPEVTTNSIPGLEL-UHFFFAOYSA-N
Synonym Diethyl Phenacylphosphonate, (2-Oxo-2-phenylethyl)phosphonic Acid Diethyl Ester
PubChem CID 372803
IUPAC Name diethyl (2-oxo-2-phenylethyl)phosphonate
SMILES CCOP(=O)(CC(=O)C1=CC=CC=C1)OCC

Specifications

Color Yellow
Boiling Point 193°C
Quantity 5 g
Formula Weight 256.24
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (2-Oxo-2-phenylethyl)phosphonate