missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (2-Methylbenzyl)phosphonate 98.0+%, TCI America™

Catalog No. p-7127668
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33275G 5 g
D332725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33275G Supplier TCI America Supplier No. D33275G

Chemical Identifiers

CAS 62778-16-9
Molecular Formula C12H19O3P
Molecular Weight (g/mol) 242.255
InChI Key SAVIMLRIKAZZCZ-UHFFFAOYSA-N
Synonym 2-(Diethylphosphonomethyl)toluene, (2-Methylbenzyl)phosphonic Acid Diethyl Ester
PubChem CID 112945
IUPAC Name 1-(diethoxyphosphorylmethyl)-2-methylbenzene
SMILES CCOP(=O)(CC1=CC=CC=C1C)OCC

Specifications

Color Colorless
Quantity 5 g
Formula Weight 242.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (2-Methylbenzyl)phosphonate