missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 2-Ethyl-2-methylmalonate 97.0+%, TCI America™
$78.29 - $393.32
Chemical Identifiers
| CAS | 2049-70-9 |
|---|---|
| Molecular Formula | C10H18O4 |
| Molecular Weight (g/mol) | 202.25 |
| MDL Number | MFCD02258541 |
| InChI Key | ODRGILDUWDVBJX-UHFFFAOYSA-N |
| Synonym | 2-Ethyl-2-methylmalonic Acid Diethyl Ester |
| PubChem CID | 16293 |
| IUPAC Name | 1,3-diethyl 2-ethyl-2-methylpropanedioate |
| SMILES | CCOC(=O)C(C)(CC)C(=O)OCC |
Chemical Identifiers
| 2049-70-9 | |
| 202.25 | |
| ODRGILDUWDVBJX-UHFFFAOYSA-N | |
| 16293 | |
| CCOC(=O)C(C)(CC)C(=O)OCC |
| C10H18O4 | |
| MFCD02258541 | |
| 2-Ethyl-2-methylmalonic Acid Diethyl Ester | |
| 1,3-diethyl 2-ethyl-2-methylpropanedioate |
Specifications
| 2049-70-9 | |
| 79°C | |
| MFCD02258541 | |
| 2-Ethyl-2-methylmalonic Acid Diethyl Ester | |
| CCOC(=O)C(C)(CC)C(=O)OCC | |
| 202.25 | |
| 202.25 | |
| Liquid |
| Yellow | |
| C10H18O4 | |
| 5 g | |
| ODRGILDUWDVBJX-UHFFFAOYSA-N | |
| 1,3-diethyl 2-ethyl-2-methylpropanedioate | |
| 16293 | |
| ≥97.0% (GC) | |
| Diethyl 2-Ethyl-2-methylmalonate |
Safety and Handling
EINECSNumber : (2)-0861&(2)-0879
TSCA : No