missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-Bromo-2-methylmalonate 98.0+%, TCI America™

Catalog No. D436825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D436825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D436825G Supplier TCI America Supplier No. D436825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29263-94-3
Molecular Formula C8H13BrO4
Molecular Weight (g/mol) 253.09
MDL Number MFCD00009125
InChI Key CSLQAXTUGPUBCW-UHFFFAOYSA-N
Synonym diethyl 2-bromo-2-methylmalonate, diethyl bromomethylmalonate, diethyl methylbromomalonate, 2-bromo-2-methylmalonic acid diethyl ester, 1,3-diethyl 2-bromo-2-methylpropanedioate, 2-bromo-2-methylpropandioic acid diethyl ester, diethyl bromo methyl malonate, acmc-20al99, cslqaxtugpubcw-uhfffaoysa, diethyl-2-bromo-2-methyl-malonate
PubChem CID 94944
IUPAC Name 1,3-diethyl 2-bromo-2-methylpropanedioate
SMILES CCOC(=O)C(C)(Br)C(=O)OCC

Specifications

Color Yellow
Boiling Point 59°C
Quantity 25 g
UN Number 3265
Formula Weight 253.09
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Diethyl 2-Bromo-2-methylmalonate