missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-bromo-2-methylmalonate, 98%

Catalog No. p-7043395
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0057514 25 g
AAL0057522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0057514 Supplier Thermo Scientific Chemicals Supplier No. L0057514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 29263-94-3 | C8H13BrO4 | 253.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29263-94-3
Molecular Formula C8H13BrO4
Molecular Weight (g/mol) 253.09
MDL Number MFCD00009125
InChI Key CSLQAXTUGPUBCW-UHFFFAOYSA-N
Synonym diethyl 2-bromo-2-methylmalonate, diethyl bromomethylmalonate, diethyl methylbromomalonate, 2-bromo-2-methylmalonic acid diethyl ester, 1,3-diethyl 2-bromo-2-methylpropanedioate, 2-bromo-2-methylpropandioic acid diethyl ester, diethyl bromo methyl malonate, acmc-20al99, cslqaxtugpubcw-uhfffaoysa, diethyl-2-bromo-2-methyl-malonate
PubChem CID 94944
IUPAC Name diethyl 2-bromo-2-methylpropanedioate
SMILES CCOC(=O)C(C)(Br)C(=O)OCC

Specifications

Density 1.323
Boiling Point 105°C to 106°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.449
Quantity 25 g
UN Number UN3265
Beilstein 1779673
Formula Weight 253.1
Percent Purity 98%
Chemical Name or Material Diethyl 2-bromo-2-methylmalonate
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 249-541-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only