missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2,6-Pyridinedicarboxylate 98.0+%, TCI America™

Catalog No. P124025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P124025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P124025G Supplier TCI America Supplier No. P124025G
Only null left

Chemical Identifiers

CAS 15658-60-3
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00033792
InChI Key KTOBUCHVPBPHMK-UHFFFAOYSA-N
Synonym Diethyl Dipicolinate, 2,6-Pyridinedicarboxylic Acid Diethyl Ester, Dipicolinic Acid Diethyl Ester
PubChem CID 254510
IUPAC Name 2,6-diethyl pyridine-2,6-dicarboxylate
SMILES CCOC(=O)C1=CC=CC(=N1)C(=O)OCC

Specifications

Melting Point 44°C
Boiling Point 188°C
Quantity 25 g
Formula Weight 223.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl 2,6-Pyridinedicarboxylate