missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2,5-Dioxahexanedioate 98.0+%, TCI America™

Catalog No. D245625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D245625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D245625G Supplier TCI America Supplier No. D245625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35466-87-6
Molecular Formula C8H14O6
Molecular Weight (g/mol) 206.194
MDL Number MFCD00460005
InChI Key MYNUAGYBVSQRFN-UHFFFAOYSA-N
Synonym 2,5-Dioxahexanedioic Acid Diethyl Ester
PubChem CID 13172823
IUPAC Name 2-ethoxycarbonyloxyethyl ethyl carbonate
SMILES CCOC(=O)OCCOC(=O)OCC

Specifications

Color Yellow
Boiling Point 227°C
Quantity 25 g
Formula Weight 206.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl 2,5-Dioxahexanedioate