missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 2,5-Dibromoadipate 98.0+%, TCI America™
Supplier: TCI America D396525G
Specifications
| Diethyl 2,5-Dibromoadipate | |
| 66°C | |
| 134°C | |
| MFCD00075379 | |
| 3261 | |
| UBCNJHBDCUBIPB-UHFFFAOYNA-N | |
| 1,6-diethyl 2,5-dibromohexanedioate | |
| 241588 | |
| ≥98.0% (GC) |
| 869-10-3 | |
| White-Yellow | |
| C10H16Br2O4 | |
| 25 g | |
| diethyl 2,5-dibromoadipate, diethyl meso-2,5-dibromoadipate, 1,6-diethyl 2,5-dibromohexanedioate, 2,5-dibromoadipic acid diethyl ester, hexanedioic acid, 2,5-dibromo-, diethyl ester, diethyl2,5-dibromohexanedioate, 2,5-dibromohexanedioic acid diethyl ester, acmc-1bjoi, diethylmeso-dibromoadipate, diethylmeso-2,5-dibromoadipate | |
| CCOC(=O)C(Br)CCC(Br)C(=O)OCC | |
| 360.04 | |
| 360.04 | |
| Crystalline Powder |
Chemical Identifiers
| 869-10-3 | |
| 360.04 | |
| UBCNJHBDCUBIPB-UHFFFAOYNA-N | |
| 241588 | |
| CCOC(=O)C(Br)CCC(Br)C(=O)OCC |
| C10H16Br2O4 | |
| MFCD00075379 | |
| diethyl 2,5-dibromoadipate, diethyl meso-2,5-dibromoadipate, 1,6-diethyl 2,5-dibromohexanedioate, 2,5-dibromoadipic acid diethyl ester, hexanedioic acid, 2,5-dibromo-, diethyl ester, diethyl2,5-dibromohexanedioate, 2,5-dibromohexanedioic acid diethyl ester, acmc-1bjoi, diethylmeso-dibromoadipate, diethylmeso-2,5-dibromoadipate | |
| 1,6-diethyl 2,5-dibromohexanedioate |
Safety and Handling
EINECSNumber : (2)-4013
TSCA : No