missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (2,4-dichlorophenyl)malonate, 95%

Catalog No. aah26330me
Change view
Click to view available options
Quantity:
500 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26330ME 500 mg
AAH2633027 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH26330ME Supplier Thermo Scientific Chemicals Supplier No. H26330ME

CAS: 111544-93-5 | C13H14Cl2O4 | 305.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111544-93-5
Molecular Formula C13H14Cl2O4
Molecular Weight (g/mol) 305.15
MDL Number MFCD00831103
InChI Key FIGCCTUCURBNIF-UHFFFAOYSA-N
Synonym diethyl 2-2,4-dichlorophenyl malonate, diethyl 2,4-dichlorophenyl malonate, 1,3-diethyl 2-2,4-dichlorophenyl propanedioate, diethyl 2-2,4-dichlorophenyl propanedioate, diethyl 2,4-dichlorophenyl propanedioate, diethyl 2,4-dichlorophenylmalonate, 2-2,4-dichloro-phenyl-malonic acid diethyl ester, propanedioic acid,2-2,4-dichlorophenyl-,1,3-diethyl ester
PubChem CID 2736112
IUPAC Name diethyl 2-(2,4-dichlorophenyl)propanedioate
SMILES CCOC(=O)C(C(=O)OCC)C1=CC=C(Cl)C=C1Cl

Specifications

Quantity 500 mg
Formula Weight 305.16
Percent Purity 95%
Chemical Name or Material Diethyl (2,4-dichlorophenyl)malonate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only