Learn More
Diethyl 2,2-diethoxyethylphosphonate, 96%
CAS: 7598-61-0 | C10H23O5P | 254.263 g/mol
Supplier: Thermo Scientific Chemicals A1246130
Description
Diethyl 2,2-diethoxyethylphosphonate is used reactant for synthesis of α,β-alkenal derivatives by two-carbon homologation, lower rim-phosphonylated rexorcinol calix[4]arenes by condensation reactions, α-Phosphovinyl radicals via a radical trapping sequence, inhibitors of reverse transcriptase via 1,3-dipolar cycloadditions and for Friedel-Crafts reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl 2,2-diethoxyethylphosphonate is used reactant for synthesis of α,β-alkenal derivatives by two-carbon homologation, lower rim-phosphonylated rexorcinol calix[4]arenes by condensation reactions, α-Phosphovinyl radicals via a radical trapping sequence, inhibitors of reverse transcriptase via 1,3-dipolar cycloadditions and for Friedel-Crafts reactions.
Solubility
Soluble in chloroform and dichloromethane. Not miscible or difficult to mix in water.
Notes
Keep container tightly sealed. It is hygroscopic in nature. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.
Specifications
| Diethyl 2,2-diethoxyethylphosphonate | |
| 1.052 | |
| >110°C (230°F) | |
| 1.4278 | |
| 250 g | |
| Hygroscopic | |
| Soluble in chloroform and dichloromethane. Not miscible or difficult to mix in water. | |
| CCOC(CP(=O)(OCC)OCC)OCC | |
| 254.263 | |
| 254.26 |
| 7598-61-0 | |
| 101°C to 103°C (8 mmHg) | |
| C10H23O5P | |
| MFCD00009236 | |
| 1785614 | |
| diethyl 2,2-diethoxyethylphosphonate, diethyl 2,2-diethoxyethyl phosphonate, diethyl2,2-diethoxyethylphosphonate, 2,2-diethoxyethylphosphonic acid diethyl ester, phosphonic acid, 2,2-diethoxyethyl-, diethyl ester, 2,2-diethoxyethyl diethoxyphosphino-1-one, diethyl phosphonacetaldehyde diethyl acetal, diethylphosphonoacetaldehyde diethylacetal, 2-diethoxyphosphoryl-1,1-diethoxy-ethane | |
| LUQYELQXRPNKRY-UHFFFAOYSA-N | |
| 2-diethoxyphosphoryl-1,1-diethoxyethane | |
| 82071 | |
| 96% |
Chemical Identifiers
| 7598-61-0 | |
| 254.263 | |
| LUQYELQXRPNKRY-UHFFFAOYSA-N | |
| 82071 | |
| CCOC(CP(=O)(OCC)OCC)OCC |
| C10H23O5P | |
| MFCD00009236 | |
| diethyl 2,2-diethoxyethylphosphonate, diethyl 2,2-diethoxyethyl phosphonate, diethyl2,2-diethoxyethylphosphonate, 2,2-diethoxyethylphosphonic acid diethyl ester, phosphonic acid, 2,2-diethoxyethyl-, diethyl ester, 2,2-diethoxyethyl diethoxyphosphino-1-one, diethyl phosphonacetaldehyde diethyl acetal, diethylphosphonoacetaldehyde diethylacetal, 2-diethoxyphosphoryl-1,1-diethoxy-ethane | |
| 2-diethoxyphosphoryl-1,1-diethoxyethane |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 231-504-0
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only