Learn More
Diethyl 12-mercaptododecylphosphonate, 95%
CAS: 1049677-30-6 | C16H35O3PS | 338.49 g/mol
$264.79 - $804.97
Chemical Identifiers
| CAS | 1049677-30-6 |
|---|---|
| Molecular Formula | C16H35O3PS |
| Molecular Weight (g/mol) | 338.49 |
| MDL Number | MFCD11046117 |
| InChI Key | JAVFXEXYSKLPHD-UHFFFAOYSA-N |
| Synonym | diethyl 12-mercaptododecylphosphonate, diethyl 12-sulfanyldodecylphosphonate, 12-mercaptododecylphosphonic acid diethyl ester |
| PubChem CID | 45158995 |
| IUPAC Name | 12-diethoxyphosphoryldodecane-1-thiol |
| SMILES | CCOP(=O)(CCCCCCCCCCCCS)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH33935MD
|
Thermo Scientific Chemicals
H33935MD |
250 mg |
Each for $264.79
|
|
|||||
|
AAH3393503
|
Thermo Scientific Chemicals
H3393503 |
1 g |
Each for $804.97
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1049677-30-6 | |
| 338.49 | |
| JAVFXEXYSKLPHD-UHFFFAOYSA-N | |
| 45158995 | |
| CCOP(=O)(CCCCCCCCCCCCS)OCC |
| C16H35O3PS | |
| MFCD11046117 | |
| diethyl 12-mercaptododecylphosphonate, diethyl 12-sulfanyldodecylphosphonate, 12-mercaptododecylphosphonic acid diethyl ester | |
| 12-diethoxyphosphoryldodecane-1-thiol |
Specifications
| 1049677-30-6 | |
| HS(CH2)12P(O)(OCH2CH3)2 | |
| MFCD11046117 | |
| Air Sensitive | |
| JAVFXEXYSKLPHD-UHFFFAOYSA-N | |
| 12-diethoxyphosphoryldodecane-1-thiol | |
| 45158995 | |
| 95% |
| C16H35O3PS | |
| 1.4610-1.4650 | |
| 250 mg | |
| diethyl 12-mercaptododecylphosphonate, diethyl 12-sulfanyldodecylphosphonate, 12-mercaptododecylphosphonic acid diethyl ester | |
| CCOP(=O)(CCCCCCCCCCCCS)OCC | |
| 338.49 | |
| 338.49 | |
| Diethyl 12-mercaptododecylphosphonate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only